Anti-HLA DR + HLA DP Mouse Monoclonal Antibody [Clone: HL-38] (PE (Phycoerythrin))
Lieferant: ANTIBODIES.COM
A85927-100
ANTIA85927-100EA 345 EUR
ANTIA85927-100
Anti-HLA DR + HLA DP Mouse Monoclonal Antibody [Clone: HL-38] (PE (Phycoerythrin))
Antikörper
Mouse monoclonal [HL-38] antibody to HLA DR + HLA DP (PE) for flow cytometry with samples derived from human and porcine.
  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 20 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: HLA DR + HLA DP
Clonality: Monoclonal
Clone: HL-38
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Porcine