Anti-HLA DR Mouse Monoclonal Antibody [Clone: MEM-267]

Lieferant: ANTIBODIES.COM
A85933-100
ANTIA85933-100EA 265 EUR
ANTIA85933-100
Anti-HLA DR Mouse Monoclonal Antibody [Clone: MEM-267]
Antikörper
Mouse monoclonal [MEM-267] antibody to HLA DR for WB, flow cytometry and ELISA with samples derived from human.

  • Validated applications: WB, Flow cytometry, ELISA

Type: Primary
Antigen: HLA DR
Clonality: Monoclonal
Clone: MEM-267
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR