Anti-LAT Mouse Monoclonal Antibody [Clone: LAT-01]

Lieferant: ANTIBODIES.COM
A86328-100
ANTIA86328-100EA 285 EUR
ANTIA86328-100
Anti-LAT Mouse Monoclonal Antibody [Clone: LAT-01]
Antikörper
Mouse monoclonal [LAT-01] antibody to LAT for WB, IP and Flow Cytometry with samples derived from Human.

  • Validated applications: WB, IP, Flow cytometry
  • Recommended dilutions: WB: 1 to 2 µg/ml

Type: Primary
Antigen: LAT
Clonality: Monoclonal
Clone: LAT-01
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR