Anti-Thyroid Hormone Receptor beta 1 Rabbit Polyclonal Antibody

Lieferant: ANTIBODIES.COM
A94592-100
ANTIA94592-100EA 475 EUR
ANTIA94592-100
Anti-Thyroid Hormone Receptor beta 1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody to TR-beta1 (phospho Ser142) for WB and ELISA with samples derived from Human, Mouse and Rat.

  • Validated applications: WB, ELISA
  • Recommended dilutions: IHC: 1:50 to 1:100, ELISA: 1:1000

Type: Primary
Antigen: Thyroid Hormone Receptor beta 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR