Anti-Aquaporin 2 Rabbit Polyclonal Antibody

Lieferant: ANTIBODIES.COM
A95552-100
ANTIA95552-100EA 475 EUR
ANTIA95552-100
Anti-Aquaporin 2 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody to Aquaporin 2 for IF and ELISA with samples derived from Human, Mouse and Rat.

  • Validated applications: IF, ELISA
  • Recommended dilutions: IF: 1:100 to 1:500, ELISA: 1:10000

Type: Primary
Antigen: Aquaporin 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR