Anti-Epsin 2 Rabbit Polyclonal Antibody

Lieferant: ANTIBODIES.COM
A98166-100
ANTIA98166-100EA 475 EUR
ANTIA98166-100
Anti-Epsin 2 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody to EPN2 for WB and ELISA with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:20000

Type: Primary
Antigen: Epsin 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR