Anti-MPS1/TTK Rabbit Polyclonal Antibody

Lieferant: ANTIBODIES.COM
A98424-100
ANTIA98424-100EA 475 EUR
ANTIA98424-100
Anti-MPS1/TTK Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody to TTK for WB, IHC and ELISA with samples derived from human and mouse.

  • Validated applications: WB, IHC, ELISA
  • Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:10000

Type: Primary
Antigen: MPS1/TTK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR