Anti-LAT Rabbit Polyclonal Antibody

Lieferant: ANTIBODIES.COM
A98493-100
ANTIA98493-100EA 475 EUR
ANTIA98493-100
Anti-LAT Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody to LAT for WB and ELISA with samples derived from human, mouse and rat.

  • Validated applications: WB, ELISA
  • Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:5000

Type: Primary
Antigen: LAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR