Anti-MOBKL2B Rabbit Polyclonal Antibody

Lieferant: ANTIBODIES.COM
A98604-100
ANTIA98604-100EA 475 EUR
ANTIA98604-100
Anti-MOBKL2B Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody to MOBKL2B for WB and ELISA with samples derived from human and mouse.

  • Validated applications: WB, ELISA
  • Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:1000

Type: Primary
Antigen: MOBKL2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR