Anti-PIK3R5 Rabbit Polyclonal Antibody

Lieferant: ANTIBODIES.COM
A98628-100
ANTIA98628-100EA 475 EUR
ANTIA98628-100
Anti-PIK3R5 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody to PIK3R5 for IHC and ELISA with samples derived from human and mouse.

  • Validated applications: IHC, ELISA
  • Recommended dilutions: IHC: 1:50 to 1:100, ELISA: 1:20000

Type: Primary
Antigen: PIK3R5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR