Anti-OR5L1 + OR5L2 Rabbit Polyclonal Antibody

Lieferant: ANTIBODIES.COM
A98684-100
ANTIA98684-100EA 475 EUR
ANTIA98684-100
Anti-OR5L1 + OR5L2 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody to OR5L1 + OR5L2 for IF and ELISA with samples derived from human.

  • Validated applications: IF, ELISA
  • Recommended dilutions: IF: 1:100 to 1:500, ELISA: 1:5000

Type: Primary
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR