Anti-OR2L5 Rabbit Polyclonal Antibody

Lieferant: ANTIBODIES.COM
A99800-100
ANTIA99800-100EA 475 EUR
ANTIA99800-100
Anti-OR2L5 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody to OR2L5 for WB, IF and ELISA with samples derived from human.

  • Validated applications: WB, IF, ELISA
  • Recommended dilutions: WB: 1:500 to 1:1000, IF: 1:100 to 1:500, ELISA: 1:20000

Type: Primary
Antigen: OR2L5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR