Anti-IKBKG Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP30005_T100
AVIVARP30005_T100EA 349 EUR
AVIVARP30005_T100
Anti-IKBKG Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IKBKG (inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR