Anti-SCRT1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP30087_P050
AVIVARP30087_P050EA 401 EUR
AVIVARP30087_P050
Anti-SCRT1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SCRT1 (scratch family zinc finger 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Rat