Anti-CHEK2 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP30335_P050
AVIVARP30335_P050EA 401 EUR
AVIVARP30335_P050
Anti-CHEK2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CHEK2 (checkpoint kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR