Anti-PBX1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP30948_P050
AVIVARP30948_P050EA 401 EUR
AVIVARP30948_P050
Anti-PBX1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PBX1 (pre-B-cell leukemia homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Human, Mouse, Pig, Rat