Anti-TAF12 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP31327_P050
AVIVARP31327_P050EA 401 EUR
AVIVARP31327_P050
Anti-TAF12 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TAF12 (TAF12 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 20kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR