Anti-ZC3H7B Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP31808_P050
AVIVARP31808_P050EA 401 EUR
AVIVARP31808_P050
Anti-ZC3H7B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZC3H7B (zinc finger CCCH-type containing 7B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR