Anti-ABT1 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP31915_P050
AVIVARP31915_P050EA 401 EUR
AVIVARP31915_P050
Anti-ABT1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ABT1 (activator of basal transcription 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR