Anti-NFIA Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP32039_P050
AVIVARP32039_P050EA 401 EUR
AVIVARP32039_P050
Anti-NFIA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NFIA (nuclear factor I/A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen