Anti-PAWR Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP32062_P050
AVIVARP32062_P050EA 401 EUR
AVIVARP32062_P050
Anti-PAWR Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PAWR (PRKC, apoptosis, WT1, regulator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen