Anti-TFDP3 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP32159_P050
AVIVARP32159_P050EA 401 EUR
AVIVARP32159_P050
Anti-TFDP3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TFDP3 (transcription factor Dp family, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human