Anti-ATF6B Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP32167_T100
AVIVARP32167_T100EA 349 EUR
AVIVARP32167_T100
Anti-ATF6B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ATF6B (activating transcription factor 6 beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR