Anti-IRX2 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP32188_T100
AVIVARP32188_T100EA 349 EUR
AVIVARP32188_T100
Anti-IRX2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IRX2 (iroquois homeobox 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen