Anti-ZNF800 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP32203_P050
AVIVARP32203_P050EA 401 EUR
AVIVARP32203_P050
Anti-ZNF800 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZNF800 (zinc finger protein 800)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen