Anti-ZNF280B Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP32237_P050
AVIVARP32237_P050EA 401 EUR
AVIVARP32237_P050
Anti-ZNF280B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZNF280B (zinc finger protein 280B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen