Anti-ASCL1 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP32355_T100
AVIVARP32355_T100EA 349 EUR
AVIVARP32355_T100
Anti-ASCL1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ASCL1 (achaete-scute family bHLH transcription factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Guinea pig, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR