Anti-SEC14L2 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP32518_P050
AVIVARP32518_P050EA 401 EUR
AVIVARP32518_P050
Anti-SEC14L2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SEC14L2 (SEC14-like 2 (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR