Anti-CCRN4L Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP32536_T100
AVIVARP32536_T100EA 349 EUR
AVIVARP32536_T100
Anti-CCRN4L Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CCRN4L (CCR4 carbon catabolite repression 4-like (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR