Anti-ACSL1 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP32784_P050
AVIVARP32784_P050EA 401 EUR
AVIVARP32784_P050
Anti-ACSL1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ACSL1 (acyl-CoA synthetase long-chain family member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR