Anti-DUSP12 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP32856_P050
AVIVARP32856_P050EA 401 EUR
AVIVARP32856_P050
Anti-DUSP12 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DUSP12 (dual specificity phosphatase 12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen