Anti-HIF1AN Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP32904_P050
AVIVARP32904_P050EA 401 EUR
AVIVARP32904_P050
Anti-HIF1AN Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HIF1AN (hypoxia inducible factor 1, alpha subunit inhibitor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen