Anti-FOXD1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP32973_P050
AVIVARP32973_P050EA 401 EUR
AVIVARP32973_P050
Anti-FOXD1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FOXD1 (forkhead box D1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat