Anti-SHOX2 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP33286_P050 ARP33286_T100
AVIVARP33286_P050EA 401 EUR
AVIVARP33286_P050 AVIVARP33286_T100
Anti-SHOX2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SHOX2 (short stature homeobox 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR