Anti-SP3 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP33336_P050
AVIVARP33336_P050EA 401 EUR
AVIVARP33336_P050
Anti-SP3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SP3 (Sp3 transcription factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR