Anti-TCF7L2 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP33414_P050
AVIVARP33414_P050EA 401 EUR
AVIVARP33414_P050
Anti-TCF7L2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TCF7L2 (transcription factor 7-like 2 (T-cell specific, HMG-box))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen