Anti-ZNF236 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP33556_P050
AVIVARP33556_P050EA 401 EUR
AVIVARP33556_P050
Anti-ZNF236 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZNF236 (zinc finger protein 236)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen