Anti-ZNF768 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP33583_T100
AVIVARP33583_T100EA 349 EUR
AVIVARP33583_T100
Anti-ZNF768 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZNF768 (zinc finger protein 768)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Human, Mouse, Rat