Anti-MLL4 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP33704_T100
AVIVARP33704_T100EA 349 EUR
AVIVARP33704_T100
Anti-MLL4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MLL4
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat