Anti-FABP4 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP33794_P050
AVIVARP33794_P050EA 401 EUR
AVIVARP33794_P050
Anti-FABP4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FABP4 (fatty acid binding protein 4, adipocyte)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen