Anti-ACADL Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP33856_P050
AVIVARP33856_P050EA 401 EUR
AVIVARP33856_P050
Anti-ACADL Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ACADL (acyl-CoA dehydrogenase, long chain)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen