Anti-PHYHIP Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP33878_T100
AVIVARP33878_T100EA 349 EUR
AVIVARP33878_T100
Anti-PHYHIP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PHYHIP (phytanoyl-CoA 2-hydroxylase interacting protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen