Anti-KIF22 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP33919_P050
AVIVARP33919_P050EA 401 EUR
AVIVARP33919_P050
Anti-KIF22 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KIF22 (kinesin family member 22)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Guinea pig, Human, Mouse, Rat