Anti-RALY Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP34074_P050
AVIVARP34074_P050EA 401 EUR
AVIVARP34074_P050
Anti-RALY Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RALY (RALY heterogeneous nuclear ribonucleoprotein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen