Anti-PPARA Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP34113_T100
AVIVARP34113_T100EA 349 EUR
AVIVARP34113_T100
Anti-PPARA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PPARA (peroxisome proliferator-activated receptor alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen