Anti-NPM1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP34114_T100
AVIVARP34114_T100EA 349 EUR
AVIVARP34114_T100
Anti-NPM1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NPM1 (nucleophosmin (nucleolar phosphoprotein B23, numatrin))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Mouse