Anti-RNF13 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP34147_P050
AVIVARP34147_P050EA 401 EUR
AVIVARP34147_P050
Anti-RNF13 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RNF13 (ring finger protein 13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen