Anti-ZMYM4 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP34210_P050
AVIVARP34210_P050EA 401 EUR
AVIVARP34210_P050
Anti-ZMYM4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZMYM4 (zinc finger, MYM-type 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen