Anti-SERTAD1 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP34309_P050 ARP34309_T100
AVIVARP34309_P050EA 401 EUR
AVIVARP34309_P050 AVIVARP34309_T100
Anti-SERTAD1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SERTAD1 (SERTA domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR