Anti-TRIM9 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP34743_P050
AVIVARP34743_P050EA 401 EUR
AVIVARP34743_P050
Anti-TRIM9 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TRIM9 (tripartite motif containing 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen